Product Details

SNP ID
rs11559172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:129095241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGAAGGCACTTGGCTGGGGCCTG[C/A]AGGCCCCCTGGAGGATGCGGTCCGT
Phenotype
MIM: 600184 MIM: 614516
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
CRAT PubMed Links

Gene Details

Gene
CRAT
Gene Name
carnitine O-acetyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000755.3 2418 UTR 3 NP_000746.2
NM_001257363.1 2418 UTR 3 NP_001244292.1
XM_005251708.3 2418 UTR 3 XP_005251765.1
XM_017014274.1 2418 UTR 3 XP_016869763.1
XM_017014275.1 2418 UTR 3 XP_016869764.1
Gene
DOLPP1
Gene Name
dolichyldiphosphatase 1
There are no transcripts associated with this gene.

View Full Product Details