Product Details

SNP ID
rs11572839
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:216504015 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCAGTTCTTTTTAAATCCACGTTT[C/T]AGTTTTATTTCCTATTTGTGAGATA
Phenotype
MIM: 602969
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESRRG PubMed Links

Gene Details

Gene
ESRRG
Gene Name
estrogen related receptor gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134285.2 4475 UTR 3 NP_001127757.1
NM_001243505.1 4475 UTR 3 NP_001230434.1
NM_001243506.1 4475 UTR 3 NP_001230435.1
NM_001243507.1 4475 UTR 3 NP_001230436.1
NM_001243509.1 4475 UTR 3 NP_001230438.1
NM_001243510.2 4475 UTR 3 NP_001230439.1
NM_001243511.2 4475 UTR 3 NP_001230440.1
NM_001243512.1 4475 UTR 3 NP_001230441.1
NM_001243513.1 4475 UTR 3 NP_001230442.1
NM_001243514.1 4475 UTR 3 NP_001230443.1
NM_001243515.1 4475 UTR 3 NP_001230444.1
NM_001243518.1 4475 UTR 3 NP_001230447.1
NM_001243519.1 4475 UTR 3 NP_001230448.1
NM_001438.3 4475 UTR 3 NP_001429.2
NM_206594.2 4475 UTR 3 NP_996317.1
NM_206595.2 4475 UTR 3 NP_996318.1
XM_011509265.2 4475 UTR 3 XP_011507567.1
XM_011509266.2 4475 UTR 3 XP_011507568.1
XM_011509267.1 4475 UTR 3 XP_011507569.1
XM_011509268.2 4475 UTR 3 XP_011507570.1
XM_011509269.2 4475 UTR 3 XP_011507571.1
XM_011509270.1 4475 UTR 3 XP_011507572.1
XM_011509271.2 4475 UTR 3 XP_011507573.1
XM_011509274.1 4475 UTR 3 XP_011507576.1
XM_011509275.1 4475 UTR 3 XP_011507577.1
XM_011509276.1 4475 UTR 3 XP_011507578.1
XM_011509277.1 4475 UTR 3 XP_011507579.1
XM_011509278.1 4475 UTR 3 XP_011507580.1
XM_011509279.1 4475 UTR 3 XP_011507581.1
XM_011509280.2 4475 UTR 3 XP_011507582.1
XM_017000621.1 4475 UTR 3 XP_016856110.1
XM_017000622.1 4475 UTR 3 XP_016856111.1
XM_017000623.1 4475 UTR 3 XP_016856112.1
XM_017000624.1 4475 UTR 3 XP_016856113.1
XM_017000625.1 4475 UTR 3 XP_016856114.1
XM_017000626.1 4475 UTR 3 XP_016856115.1
XM_017000627.1 4475 UTR 3 XP_016856116.1
XM_017000628.1 4475 UTR 3 XP_016856117.1
XM_017000629.1 4475 UTR 3 XP_016856118.1
XM_017000630.1 4475 UTR 3 XP_016856119.1
XM_017000631.1 4475 UTR 3 XP_016856120.1
XM_017000632.1 4475 UTR 3 XP_016856121.1
XM_017000633.1 4475 UTR 3 XP_016856122.1
XM_017000634.1 4475 UTR 3 XP_016856123.1
XM_017000635.1 4475 UTR 3 XP_016856124.1
XM_017000636.1 4475 UTR 3 XP_016856125.1
XM_017000637.1 4475 UTR 3 XP_016856126.1
XM_017000638.1 4475 UTR 3 XP_016856127.1
XM_017000639.1 4475 UTR 3 XP_016856128.1
XM_017000640.1 4475 UTR 3 XP_016856129.1
XM_017000641.1 4475 UTR 3 XP_016856130.1
XM_017000642.1 4475 UTR 3 XP_016856131.1
XM_017000643.1 4475 UTR 3 XP_016856132.1
XM_017000644.1 4475 UTR 3 XP_016856133.1
XM_017000645.1 4475 UTR 3 XP_016856134.1
XM_017000646.1 4475 UTR 3 XP_016856135.1
XM_017000647.1 4475 UTR 3 XP_016856136.1
XM_017000648.1 4475 UTR 3 XP_016856137.1
XM_017000649.1 4475 UTR 3 XP_016856138.1

View Full Product Details