Product Details

SNP ID
rs11574996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:46054155 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAATAGGAATTATGAATTCTGAT[A/C]ATTTTATGGGCGAAAAATAATATCC
Phenotype
MIM: 603101 MIM: 616453
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CPB2 PubMed Links

Gene Details

Gene
CPB2
Gene Name
carboxypeptidase B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278541.1 Intron NP_001265470.1
NM_001872.4 Intron NP_001863.3
XM_017020393.1 Intron XP_016875882.1
Gene
CPB2-AS1
Gene Name
CPB2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
ZC3H13
Gene Name
zinc finger CCCH-type containing 13
There are no transcripts associated with this gene.

View Full Product Details