Product Details
- SNP ID
-
rs11593448
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:98458266 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTCCTCCTTCTAACAAAAGCATCA[C/T]TCCTGTCATTTATCCTGAGAAAGAA
- Phenotype
-
MIM: 613469
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HPSE2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs140887054] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HPSE2
- Gene Name
- heparanase 2 (inactive)
- Gene
- LOC101927278
- Gene Name
- uncharacterized LOC101927278
There are no transcripts associated with this gene.
View Full Product Details