Product Details

SNP ID
rs11596220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133396090 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAATTGGCAGGATGTGATGATTC[A/G]TTCACTGATGGCAAGAAAAAAAGTT
Phenotype
MIM: 615853
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTG1 PubMed Links

Gene Details

Gene
MTG1
Gene Name
mitochondrial ribosome associated GTPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138384.2 Intron NP_612393.2
Gene
PAOX
Gene Name
polyamine oxidase (exo-N4-amino)
There are no transcripts associated with this gene.

View Full Product Details