Product Details

SNP ID
rs11650919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4947192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGGAAAAGCAGAAGCGGAGAGG[A/G]GAAGGGAGGGGGGCGGGCGAGGCAA
Phenotype
MIM: 131370 MIM: 176610 MIM: 610431 MIM: 604165
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENO3 PubMed Links

Gene Details

Gene
ENO3
Gene Name
enolase 3
There are no transcripts associated with this gene.

Gene
PFN1
Gene Name
profilin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005022.3 Intron NP_005013.1
XM_017024761.1 Intron XP_016880250.1
Gene
RNF167
Gene Name
ring finger protein 167
There are no transcripts associated with this gene.

Gene
SLC25A11
Gene Name
solute carrier family 25 member 11
There are no transcripts associated with this gene.

View Full Product Details