Product Details

SNP ID
rs11688445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24174703 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAAACCATTCCTCTTACGTTGTC[A/G]CCCTCAATCTTACAGGTAAATGGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM228A PubMed Links

Gene Details

Gene
FAM228A
Gene Name
family with sequence similarity 228 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040710.2 Intron NP_001035800.1
Gene
FAM228B
Gene Name
family with sequence similarity 228 member B
There are no transcripts associated with this gene.

View Full Product Details