Product Details
- SNP ID
-
rs1220701
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:129611734 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGAGGCTTGGTCCCAAGTTTTTTG[G/A]GTTTCGTTGTTGTTGTTGTTTGTTT
- Phenotype
-
MIM: 613560
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C9orf50
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117337486] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C9orf50
- Gene Name
- chromosome 9 open reading frame 50
- Gene
- NTMT1
- Gene Name
- N-terminal Xaa-Pro-Lys N-methyltransferase 1
View Full Product Details