Product Details

SNP ID
rs13371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:122443881 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTATCTGAGCTATTTAAGTCTGTA[C/T]TTCAGTGTCTTCATTTTTTTAAATG
Phenotype
MIM: 614548
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERINC1 PubMed Links

Gene Details

Gene
SERINC1
Gene Name
serine incorporator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020755.3 2669 UTR 3 NP_065806.1

View Full Product Details