Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001144875.1 | 2111 | UTR 3 | NP_001138347.1 | ||
NM_001144876.1 | 2111 | UTR 3 | NP_001138348.1 | ||
NM_001308235.1 | 2111 | Intron | NP_001295164.1 | ||
NM_001308236.1 | 2111 | Intron | NP_001295165.1 | ||
NM_024872.2 | 2111 | Intron | NP_079148.2 | ||
XM_005265985.3 | 2111 | UTR 3 | XP_005266042.1 | ||
XM_005265987.1 | 2111 | UTR 3 | XP_005266044.1 | ||
XM_011534654.2 | 2111 | UTR 3 | XP_011532956.1 | ||
XM_011534655.2 | 2111 | UTR 3 | XP_011532957.1 | ||
XM_011534656.2 | 2111 | UTR 3 | XP_011532958.1 | ||
XM_011534657.2 | 2111 | UTR 3 | XP_011532959.1 | ||
XM_017009883.1 | 2111 | UTR 3 | XP_016865372.1 | ||
XM_017009884.1 | 2111 | UTR 3 | XP_016865373.1 |