Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001039887.2 | Intron | NP_001034976.2 | |||
XM_011526528.1 | Intron | XP_011524830.1 | |||
XM_011526529.2 | Intron | XP_011524831.1 | |||
XM_011526530.1 | Intron | XP_011524832.1 | |||
XM_011526531.1 | Intron | XP_011524833.1 | |||
XM_011526532.1 | Intron | XP_011524834.1 | |||
XM_011526533.2 | Intron | XP_011524835.1 | |||
XM_011526534.1 | Intron | XP_011524836.1 | |||
XM_011526535.2 | Intron | XP_011524837.1 | |||
XM_011526536.2 | Intron | XP_011524838.1 |