Product Details
- SNP ID
-
rs180812726
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:102114339 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTGAGATACCTGATGATGTACCT[C/T]TGCCAGCAGGTTGGGAGATGGCAAA
- Phenotype
-
MIM: 606608
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
YAP1
PubMed Links
Gene Details
- Gene
- YAP1
- Gene Name
- Yes associated protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001130145.2 |
905 |
Silent Mutation |
CTG,TTG |
L173L |
NP_001123617.1 |
NM_001195044.1 |
905 |
Silent Mutation |
CTG,TTG |
L173L |
NP_001181973.1 |
NM_001195045.1 |
905 |
UTR 5 |
|
|
NP_001181974.1 |
NM_001282097.1 |
905 |
Silent Mutation |
CTG,TTG |
L173L |
NP_001269026.1 |
NM_001282098.1 |
905 |
Silent Mutation |
CTG,TTG |
L173L |
NP_001269027.1 |
NM_001282099.1 |
905 |
Silent Mutation |
CTG,TTG |
L173L |
NP_001269028.1 |
NM_001282100.1 |
905 |
Silent Mutation |
CTG,TTG |
L173L |
NP_001269029.1 |
NM_001282101.1 |
905 |
Silent Mutation |
CTG,TTG |
L173L |
NP_001269030.1 |
NM_006106.4 |
905 |
Silent Mutation |
CTG,TTG |
L173L |
NP_006097.2 |
XM_005271378.3 |
905 |
Silent Mutation |
CTG,TTG |
L173L |
XP_005271435.1 |
XM_005271380.3 |
905 |
Silent Mutation |
CTG,TTG |
L173L |
XP_005271437.1 |
XM_005271381.3 |
905 |
Silent Mutation |
CTG,TTG |
L173L |
XP_005271438.1 |
XM_005271383.3 |
905 |
Silent Mutation |
CTG,TTG |
L173L |
XP_005271440.1 |
XM_011542555.2 |
905 |
UTR 5 |
|
|
XP_011540857.1 |
XM_011542556.2 |
905 |
UTR 5 |
|
|
XP_011540858.1 |
XM_017017093.1 |
905 |
UTR 5 |
|
|
XP_016872582.1 |
View Full Product Details