Product Details

SNP ID
rs180808688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:71548663 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTCAATCCACCAGAACCATGGG[C/T]TGCTGTGGCTGCTCCGGAGGCTGTG
Phenotype
MIM: 148021
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP5-8 PubMed Links

Gene Details

Gene
KRTAP5-8
Gene Name
keratin associated protein 5-8
There are no transcripts associated with this gene.

Gene
KRTAP5-9
Gene Name
keratin associated protein 5-9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005553.3 244 Silent Mutation GGC,GGT G2G NP_005544.4

View Full Product Details