Product Details

SNP ID
rs180778128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137048968 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCTGGAAGATCACGCCGCCGAA[A/G]GCGCGCTCGTCGAAGCCGTAGCCGC
Phenotype
MIM: 602012 MIM: 605798
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENTPD2 PubMed Links

Gene Details

Gene
ENTPD2
Gene Name
ectonucleoside triphosphate diphosphohydrolase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001246.3 1242 Silent Mutation GCC,GCT A396A NP_001237.1
NM_203468.2 1242 Silent Mutation GCC,GCT A419A NP_982293.1
XM_011519212.2 1242 Silent Mutation GCC,GCT A316A XP_011517514.1
Gene
NPDC1
Gene Name
neural proliferation, differentiation and control 1
There are no transcripts associated with this gene.

View Full Product Details