Product Details

SNP ID
rs181216678
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34404984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTGCCAGAGAGAGAGGCGCAGGA[C/G]GCTCAGGTAAAGGTTGTGATTGGCA
Phenotype
MIM: 172400 MIM: 615661
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GPI PubMed Links

Gene Details

Gene
GPI
Gene Name
glucose-6-phosphate isomerase
There are no transcripts associated with this gene.

Gene
PDCD2L
Gene Name
programmed cell death 2 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032346.1 363 Missense Mutation GAC,GAG D110E NP_115722.1

View Full Product Details