Product Details

SNP ID
rs181124344
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:153715646 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCGATAAGCTCCTTGTAGGCAAC[C/T]GGCTGACACTGTAGAACTATCAGAG
Phenotype
MIM: 603028
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
RNF175 PubMed Links

Gene Details

Gene
RNF175
Gene Name
ring finger protein 175
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173662.2 1104 Missense Mutation CAG,CGG Q216R NP_775933.1
XM_005262938.3 1104 Missense Mutation CAG,CGG Q216R XP_005262995.1
XM_005262939.3 1104 Missense Mutation CAG,CGG Q136R XP_005262996.1
XM_005262940.4 1104 Missense Mutation CAG,CGG Q135R XP_005262997.1
XM_011531879.2 1104 Missense Mutation CAG,CGG Q184R XP_011530181.1
XM_011531881.2 1104 Missense Mutation CAG,CGG Q176R XP_011530183.1
XM_011531882.2 1104 Missense Mutation CAG,CGG Q216R XP_011530184.1
XM_011531883.2 1104 Missense Mutation CAG,CGG Q144R XP_011530185.1
XM_017008047.1 1104 Missense Mutation CAG,CGG Q117R XP_016863536.1
Gene
TLR2
Gene Name
toll like receptor 2
There are no transcripts associated with this gene.

View Full Product Details