Product Details

SNP ID
rs181127138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36748567 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCGATGTTGAATTAGTGCTGAGC[A/G]AAGAGTAAAAGATTTCCCACATTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF850 PubMed Links

Gene Details

Gene
ZNF850
Gene Name
zinc finger protein 850
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193552.1 2536 Missense Mutation CGC,TGC R825C NP_001180481.1
NM_001267779.1 2536 Missense Mutation CGC,TGC R793C NP_001254708.1

View Full Product Details