Product Details

SNP ID
rs180896668
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:66877353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGATGAGTGCCTGCGGGAGGAA[C/G]GCCCTGACCCTGCTGAGCAGTGTCT
Phenotype
MIM: 606405
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
CACNG5 PubMed Links

Gene Details

Gene
CACNG5
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145811.2 81 Silent Mutation AAC,AAG N7K NP_665810.1

View Full Product Details