Product Details

SNP ID
rs180949263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:166198934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGACATTTTGCCTTAAGCGGTAA[C/T]GTCTATAAGCACGCTGAATGACAGT
Phenotype
MIM: 603415
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929680 PubMed Links

Gene Details

Gene
LOC101929680
Gene Name
uncharacterized LOC101929680
There are no transcripts associated with this gene.

Gene
SCN9A
Gene Name
sodium voltage-gated channel alpha subunit 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002977.3 5980 Missense Mutation CAT,CGT H1891R NP_002968.1
XM_005246757.2 5980 Missense Mutation CAT,CGT H1902R XP_005246814.1
XM_011511616.2 5980 Missense Mutation CAT,CGT H1902R XP_011509918.1
XM_011511617.2 5980 Missense Mutation CAT,CGT H1902R XP_011509919.1
XM_011511618.2 5980 Missense Mutation CAT,CGT H1891R XP_011509920.1
XM_011511619.2 5980 Intron XP_011509921.1
XM_017004668.1 5980 Missense Mutation CAT,CGT H1773R XP_016860157.1
XM_017004669.1 5980 Missense Mutation CAT,CGT H1654R XP_016860158.1

View Full Product Details