Product Details
- SNP ID
-
rs180932774
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:37866505 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCCAAGCAGTCCCTGATATGTTC[A/C]AATTCTGACCTCAGGCCTGGTTGCT
- Phenotype
-
MIM: 147264
MIM: 600172
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
INPP5B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs568256858] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- INPP5B
- Gene Name
- inositol polyphosphate-5-phosphatase B
- Gene
- MTF1
- Gene Name
- metal regulatory transcription factor 1
There are no transcripts associated with this gene.
View Full Product Details