Product Details

SNP ID
rs180987786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35995292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCGGTCATGAGCCGGCACAGCCG[A/G]CTGCAGAGGCAGGTTCTGAGCCTGT
Phenotype
MIM: 612848 MIM: 615535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SDHAF1 PubMed Links

Gene Details

Gene
SDHAF1
Gene Name
succinate dehydrogenase complex assembly factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042631.2 105 Silent Mutation CGA,CGG R6R NP_001036096.1
Gene
SYNE4
Gene Name
spectrin repeat containing nuclear envelope family member 4
There are no transcripts associated with this gene.

View Full Product Details