Product Details

SNP ID
rs181022873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:135734003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGTACTCCCGGCTGTGAGGAG[C/T]GCATCAAGTCGTGCCTCAGGGCCTG
Phenotype
MIM: 604309
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM180A PubMed Links

Gene Details

Gene
FAM180A
Gene Name
family with sequence similarity 180 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_205855.3 760 Missense Mutation CAC,CGC H165R NP_995327.1
XM_006715983.3 760 Missense Mutation CAC,CGC H165R XP_006716046.1
Gene
SLC13A4
Gene Name
solute carrier family 13 member 4
There are no transcripts associated with this gene.

View Full Product Details