Product Details

SNP ID
rs181038580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56232188 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCATTGAGTCTTTTACATCTTTG[C/T]TCCTCAAACTGTAGATGACGGGATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5T2 PubMed Links

Gene Details

Gene
OR5T2
Gene Name
olfactory receptor family 5 subfamily T member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004746.1 998 Missense Mutation AAC,AGC N333S NP_001004746.1

View Full Product Details