Product Details

SNP ID
rs180753483
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:194644608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTTTTGTCAACGATATTTTCAA[C/T]CTGCTTTGCTTTTTTATTTCTGCCT
Phenotype
MIM: 610780
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LSG1 PubMed Links

Gene Details

Gene
LSG1
Gene Name
large 60S subunit nuclear export GTPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018385.2 1405 Missense Mutation ATT,GTT I588V NP_060855.2
XM_017006796.1 1405 Missense Mutation ATT,GTT I430V XP_016862285.1

View Full Product Details