Product Details
- SNP ID
-
rs180767274
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:25161605 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCCTGCGCCGCTGCTGCCGCTGC[C/T]GCTGCTGTTGCGGCGGCCGAATCGG
- Phenotype
-
MIM: 616797
MIM: 176830
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EFR3B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201517327,rs28930368] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EFR3B
- Gene Name
- EFR3 homolog B
There are no transcripts associated with this gene.
- Gene
- POMC
- Gene Name
- proopiomelanocortin
View Full Product Details