Product Details

SNP ID
rs180707620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:78300603 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATTTTCCTTTATTTTTTTCTAGG[G/T]GTTGTTAGAGATACAAAAGGAATTA
Phenotype
MIM: 610936
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PSAT1 PubMed Links

Gene Details

Gene
PSAT1
Gene Name
phosphoserine aminotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021154.4 Intron NP_066977.1
NM_058179.3 Intron NP_478059.1

View Full Product Details