Product Details

SNP ID
rs181321816
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:5086022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGAGGACCTGGAGCAGCTCGGGGG[A/G]CTGCTGGCCGCGGCGGCAGTGGCCG
Phenotype
MIM: 613311
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LYRM4 PubMed Links

Gene Details

Gene
LYRM4
Gene Name
LYR motif containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164840.2 537 Intron NP_001158312.1
NM_001164841.2 537 Intron NP_001158313.1
NM_001318782.1 537 Intron NP_001305711.1
NM_001318783.1 537 Intron NP_001305712.1
NM_020408.5 537 Intron NP_065141.3
XM_017011083.1 537 Intron XP_016866572.1
XM_017011084.1 537 Intron XP_016866573.1
Gene
LYRM4-AS1
Gene Name
LYRM4 antisense RNA 1
There are no transcripts associated with this gene.

Gene
PPP1R3G
Gene Name
protein phosphatase 1 regulatory subunit 3G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145115.2 537 Silent Mutation GGA,GGG G179G NP_001138587.1

View Full Product Details