Product Details

SNP ID
rs181335004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55262626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGTTTCACCGATCCATGCTGTC[A/G]TCCGCTGGGCTGGAGAAACAGGTCT
Phenotype
MIM: 612939 MIM: 610875
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSPBP1 PubMed Links

Gene Details

Gene
HSPBP1
Gene Name
HSPA (Hsp70) binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130106.1 1505 Silent Mutation NP_001123578.1
NM_001297600.1 1505 Silent Mutation NP_001284529.1
NM_012267.4 1505 Silent Mutation NP_036399.3
XM_005258700.2 1505 Missense Mutation XP_005258757.1
XM_005258701.2 1505 Missense Mutation XP_005258758.1
XM_011526691.1 1505 Missense Mutation XP_011524993.1
XM_017026543.1 1505 Missense Mutation XP_016882032.1
XM_017026544.1 1505 Silent Mutation XP_016882033.1
XM_017026545.1 1505 UTR 3 XP_016882034.1
Gene
PPP6R1
Gene Name
protein phosphatase 6 regulatory subunit 1
There are no transcripts associated with this gene.

View Full Product Details