Product Details

SNP ID
rs181326809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:118380213 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCAAAGCATGATGAAGAAGACT[A/G]TGTAGAAATGAAGGAACAGTGAGTA
Phenotype
MIM: 608250 MIM: 616711
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SUDS3 PubMed Links

Gene Details

Gene
SUDS3
Gene Name
SDS3 homolog, SIN3A corepressor complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022491.2 333 Missense Mutation TAT,TGT Y65C NP_071936.2
XM_011538671.2 333 Missense Mutation TAT,TGT Y144C XP_011536973.1
XM_017019817.1 333 Intron XP_016875306.1
XM_017019818.1 333 UTR 5 XP_016875307.1
XM_017019819.1 333 UTR 5 XP_016875308.1
XM_017019820.1 333 UTR 5 XP_016875309.1
Gene
TAOK3
Gene Name
TAO kinase 3
There are no transcripts associated with this gene.

View Full Product Details