Product Details

SNP ID
rs181648829
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:160681105 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACACCTTAGGGATTCACCACCAC[A/G]CTGTGCAAGGAGGCTGAATAGGACC
Phenotype
MIM: 109530
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD48 PubMed Links

Gene Details

Gene
CD48
Gene Name
CD48 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256030.1 837 Missense Mutation NP_001242959.1
NM_001778.3 837 Intron NP_001769.2
XM_005245625.1 837 Intron XP_005245682.1
XM_011510171.1 837 Intron XP_011508473.1
XM_017002867.1 837 Intron XP_016858356.1

View Full Product Details