Product Details
- SNP ID
-
rs181619166
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:69999790 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTCATTTTTCACAGAAATTTTTGC[A/G]TAGAATTGGAAGATTCGGTGTAAGT
- Phenotype
-
MIM: 184470
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
STATH
PubMed Links
Gene Details
- Gene
- STATH
- Gene Name
- statherin
View Full Product Details