Product Details

SNP ID
rs181775983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:151337878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTCTTTTTTCCTATTGTCCTGG[A/G]ACAGAGATGTTGCAGAATTGGGGCA
Phenotype
MIM: 611318 MIM: 600515 MIM: 606380
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MED12L PubMed Links

Gene Details

Gene
MED12L
Gene Name
mediator complex subunit 12 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053002.5 1526 Intron NP_443728.3
XM_006713487.3 1526 Intron XP_006713550.1
XM_011512390.2 1526 Intron XP_011510692.1
XM_011512394.2 1526 Intron XP_011510696.1
XM_011512399.2 1526 Intron XP_011510701.1
XM_017005676.1 1526 Intron XP_016861165.1
XM_017005677.1 1526 Intron XP_016861166.1
XM_017005678.1 1526 Intron XP_016861167.1
XM_017005679.1 1526 Intron XP_016861168.1
XM_017005680.1 1526 Intron XP_016861169.1
XM_017005681.1 1526 Intron XP_016861170.1
Gene
P2RY12
Gene Name
purinergic receptor P2Y12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022788.4 1526 Missense Mutation TCC,TTC S323F NP_073625.1
NM_176876.2 1526 Missense Mutation TCC,TTC S323F NP_795345.1
XM_017007069.1 1526 Missense Mutation TCC,TTC S323F XP_016862558.1
Gene
P2RY13
Gene Name
purinergic receptor P2Y13
There are no transcripts associated with this gene.

View Full Product Details