Product Details

SNP ID
rs181729673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30427336 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGCCACCCGCTGGCCCCTGGCT[A/G]TCCGCAAGTGGACCACCACCGCGCT
Phenotype
MIM: 610235 MIM: 607558
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTFP1 PubMed Links

Gene Details

Gene
MTFP1
Gene Name
mitochondrial fission process 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003704.2 618 Intron NP_001003704.1
NM_016498.4 618 Missense Mutation ATC,GTC I121V NP_057582.2
Gene
SEC14L2
Gene Name
SEC14 like lipid binding 2
There are no transcripts associated with this gene.

View Full Product Details