Product Details

SNP ID
rs181790702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:97854660 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGAAGCCTCTCACTCTCCTCCTC[A/G]GCTTTTTCAGGAGAAGGAGCCTATT
Phenotype
MIM: 108370
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASNS PubMed Links

Gene Details

Gene
ASNS
Gene Name
asparagine synthetase (glutamine-hydrolyzing)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178075.1 1034 Intron NP_001171546.1
NM_001178076.1 1034 Intron NP_001171547.1
NM_001178077.1 1034 Silent Mutation GCC,GCT A303A NP_001171548.1
NM_001673.4 1034 Intron NP_001664.3
NM_133436.3 1034 Silent Mutation GCC,GCT A386A NP_597680.2
NM_183356.3 1034 Intron NP_899199.2

View Full Product Details