Product Details

SNP ID
rs181743036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:31337751 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACCTGTGTGCTGAGCTCGAGCTG[C/T]ACGGATGGCATCTTTGATGGCAAAG
Phenotype
MIM: 607633
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
XDH PubMed Links

Gene Details

Gene
XDH
Gene Name
xanthine dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000379.3 3920 Missense Mutation ACA,GCA T1281A NP_000370.2
XM_011533095.2 3920 Missense Mutation ACA,GCA T1280A XP_011531397.1
XM_011533096.2 3920 Intron XP_011531398.1

View Full Product Details