Product Details

SNP ID
rs181685539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:79236677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCGTTCGCGCTGGAGGAGACGCT[C/T]GAGTCGGACTGGGTGGCTGTGCGGC
Phenotype
MIM: 604279
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JMY PubMed Links

Gene Details

Gene
JMY
Gene Name
junction mediating and regulatory protein, p53 cofactor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152405.4 576 Silent Mutation CTC,CTT L9L NP_689618.4
XM_005248430.2 576 Silent Mutation CTC,CTT L9L XP_005248487.1
XM_011543155.2 576 Silent Mutation CTC,CTT L9L XP_011541457.1

View Full Product Details