Product Details

SNP ID
rs181397131
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:789225 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCCCCAGCCCCCAGGATCAAAC[A/G]GCCTAGGCTGCAGGTGGTCAAGAGG
Phenotype
MIM: 613201 MIM: 607298
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CHTF18 PubMed Links
Additional Information
For this assay, SNP(s) [rs77682584] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CHTF18
Gene Name
chromosome transmission fidelity factor 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022092.2 364 Missense Mutation CAG,CGG Q101R NP_071375.1
XM_005255470.1 364 UTR 5 XP_005255527.1
XM_005255471.3 364 Missense Mutation CAG,CGG Q129R XP_005255528.1
XM_011522572.1 364 Missense Mutation CAG,CGG Q102R XP_011520874.1
XM_011522573.1 364 Intron XP_011520875.1
XM_017023532.1 364 Missense Mutation CAG,CGG Q80R XP_016879021.1
XM_017023533.1 364 Missense Mutation CAG,CGG Q102R XP_016879022.1
XM_017023534.1 364 UTR 5 XP_016879023.1
Gene
GNG13
Gene Name
G protein subunit gamma 13
There are no transcripts associated with this gene.

Gene
RPUSD1
Gene Name
RNA pseudouridylate synthase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324086.1 364 Intron NP_001311015.1
NM_001324410.1 364 Intron NP_001311339.1
NM_001324411.1 364 Intron NP_001311340.1
NM_001324412.1 364 Intron NP_001311341.1
NM_001324413.1 364 Intron NP_001311342.1
NM_001324414.1 364 Intron NP_001311343.1
NM_001324415.1 364 Intron NP_001311344.1
NM_058192.2 364 Intron NP_478072.1

View Full Product Details