Product Details

SNP ID
rs181395905
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:46560252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTACCTCTGGAGGACTTGCGGC[A/G]TGGGGAGTCCCTTTTCTGGAGCTTG
Phenotype
MIM: 611455 MIM: 606724
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KNCN PubMed Links

Gene Details

Gene
KNCN
Gene Name
kinocilin
There are no transcripts associated with this gene.

Gene
MKNK1
Gene Name
MAP kinase interacting serine/threonine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135553.2 6605 Missense Mutation NP_001129025.1
NM_003684.5 6605 Missense Mutation NP_003675.2
NM_198973.3 6605 Intron NP_945324.1
XM_006711000.1 6605 Missense Mutation XP_006711063.1
XM_006711001.2 6605 Missense Mutation XP_006711064.1
XM_006711002.2 6605 Missense Mutation XP_006711065.1
XM_006711003.1 6605 Missense Mutation XP_006711066.1
XM_006711006.3 6605 Missense Mutation XP_006711069.1
XM_006711007.1 6605 Missense Mutation XP_006711070.1
XM_006711009.3 6605 Missense Mutation XP_006711072.1
XM_006711013.1 6605 Missense Mutation XP_006711076.1
XM_006711014.1 6605 Missense Mutation XP_006711077.1
XM_006711015.1 6605 Missense Mutation XP_006711078.1
XM_017002652.1 6605 Missense Mutation XP_016858141.1
XM_017002653.1 6605 Missense Mutation XP_016858142.1
XM_017002654.1 6605 Missense Mutation XP_016858143.1
XM_017002655.1 6605 Missense Mutation XP_016858144.1
XM_017002656.1 6605 Missense Mutation XP_016858145.1
XM_017002657.1 6605 Missense Mutation XP_016858146.1
XM_017002658.1 6605 Intron XP_016858147.1
XM_017002659.1 6605 Intron XP_016858148.1
XM_017002660.1 6605 Intron XP_016858149.1
XM_017002661.1 6605 Intron XP_016858150.1
XM_017002662.1 6605 Intron XP_016858151.1
XM_017002663.1 6605 Intron XP_016858152.1
XM_017002664.1 6605 Missense Mutation XP_016858153.1
XM_017002665.1 6605 Intron XP_016858154.1
XM_017002666.1 6605 Intron XP_016858155.1
XM_017002667.1 6605 Intron XP_016858156.1
XM_017002668.1 6605 Intron XP_016858157.1
Gene
MKNK1-AS1
Gene Name
MKNK1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details