Product Details
- SNP ID
-
rs181485765
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:70055506 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTTGTGTGTATGCAGGAAAAGCA[A/T]CATTCACATCGAGAATTTCCATTTT
- Phenotype
-
MIM: 142701
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HTN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61744465] are located under a probe and SNP(s) [rs13353559] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HTN1
- Gene Name
- histatin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002159.3 |
205 |
Missense Mutation |
CAA,CAT |
Q37H |
NP_002150.1 |
View Full Product Details