Product Details
- SNP ID
-
rs181553624
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:26139665 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCTGGGAAATTATGAATATGATGG[A/G]TGTATATGCCGATAGGAAGCACTTC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MAGEB18
PubMed Links
Gene Details
- Gene
- MAGEB18
- Gene Name
- MAGE family member B18
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173699.3 |
867 |
Missense Mutation |
GAT,GGT |
D227G |
NP_775970.1 |
View Full Product Details