Product Details

SNP ID
rs181531193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184033045 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTCAACATCTCCTTCACCTTGT[C/T]TGCCATCTGGAAGTTGTAAGTCCTC
Phenotype
MIM: 610122
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HTR3D PubMed Links

Gene Details

Gene
HTR3D
Gene Name
5-hydroxytryptamine receptor 3D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145143.1 215 Intron NP_001138615.1
NM_001163646.1 215 Missense Mutation TCT,TTT S72F NP_001157118.1
NM_182537.2 215 Intron NP_872343.2
XM_017005854.1 215 Intron XP_016861343.1

View Full Product Details