Product Details

SNP ID
rs181260849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112978688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGTGGAGGAAGGGCCGTCTATC[A/C]ATCAAGTGAATCAAATGCTGTTGGG
Phenotype
MIM: 605542
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IL36G PubMed Links

Gene Details

Gene
IL36G
Gene Name
interleukin 36, gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278568.1 129 Missense Mutation CAA,CCA Q17P NP_001265497.1
NM_019618.3 129 Missense Mutation CAA,CCA Q17P NP_062564.1

View Full Product Details