Product Details
- SNP ID
-
rs181308670
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:36940348 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCATCTCCCCTAGCGACCTGGTG[A/C]GGAGAGCGCGGAGCAAAGTGTTTCA
- Phenotype
-
MIM: 608463
MIM: 601999
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AATF
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs536612068] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AATF
- Gene Name
- apoptosis antagonizing transcription factor
There are no transcripts associated with this gene.
- Gene
- LHX1
- Gene Name
- LIM homeobox 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005568.4 |
952 |
Silent Mutation |
AGG,CGG |
R77R |
NP_005559.2 |
- Gene
- LOC102723471
- Gene Name
- uncharacterized LOC102723471
There are no transcripts associated with this gene.
View Full Product Details