Product Details
- SNP ID
-
rs181316271
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:34961630 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGAGTGATAAATCAGGAAGAATTG[A/C]GGCGGTTAATGAAGGAGAAGCAGCG
- Phenotype
-
MIM: 610730
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CCT6B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77056680] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCT6B
- Gene Name
- chaperonin containing TCP1 subunit 6B
- Gene
- ZNF830
- Gene Name
- zinc finger protein 830
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_052857.3 |
101 |
Silent Mutation |
AGG,CGG |
R22R |
NP_443089.3 |
View Full Product Details