Product Details

SNP ID
rs181910740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63832989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCAGTTCCTGCCTTCGCTGCTG[C/T]ACCTGGAGAAGGGAGAAACCAAGTG
Phenotype
MIM: 601681 MIM: 601736
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FTSJ3 PubMed Links

Gene Details

Gene
FTSJ3
Gene Name
FtsJ homolog 3
There are no transcripts associated with this gene.

Gene
PSMC5
Gene Name
proteasome 26S subunit, ATPase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199163.1 1409 Intron NP_001186092.1
NM_002805.5 1409 Intron NP_002796.4
Gene
SMARCD2
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098426.1 1409 Silent Mutation GTA,GTG V515V NP_001091896.1
XM_005257604.2 1409 Silent Mutation GTA,GTG V440V XP_005257661.2
XM_017024967.1 1409 Silent Mutation GTA,GTG V467V XP_016880456.1

View Full Product Details