Product Details

SNP ID
rs182287744
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37571849 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCAACCCTGAAACCTTGCTCAC[A/C]CATCAGTGCCATCGGCGATGCTGCC
Phenotype
MIM: 606084 MIM: 150571
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDC42EP1 PubMed Links

Gene Details

Gene
CDC42EP1
Gene Name
CDC42 effector protein 1
There are no transcripts associated with this gene.

Gene
LGALS2
Gene Name
galectin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006498.2 201 Missense Mutation GGC,GTC G30V NP_006489.1

View Full Product Details