Product Details

SNP ID
rs182310862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:77040508 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGGGCATGTGTCTGGAGCGCGC[C/T]GGCCTGGCGCCCAGCGGCCACGAGG
Phenotype
MIM: 615315
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B3GNT6 PubMed Links

Gene Details

Gene
B3GNT6
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138706.4 1095 Silent Mutation GCC,GCT A319A NP_619651.3

View Full Product Details