Product Details
- SNP ID
-
rs182273528
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
10
- Location
-
Chr.1:221332315 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGCAATTTATGACACATATAACA[A/G]TATGACTGAATCTTGAGGATATTAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C1orf140
PubMed Links
Gene Details
- Gene
- C1orf140
- Gene Name
- uncharacterized LOC400804
There are no transcripts associated with this gene.
View Full Product Details