Product Details

SNP ID
rs181949767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28985436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCAGATGGAGGAGGCCATCCTG[G/T]TCCCCTGCGTGCTGGGGCTCCTGCT
Phenotype
MIM: 602354 MIM: 612583
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LAT PubMed Links

Gene Details

Gene
LAT
Gene Name
linker for activation of T-cells
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014987.1 371 Missense Mutation GTC,TTC V7F NP_001014987.1
NM_001014988.1 371 Missense Mutation GTC,TTC V7F NP_001014988.1
NM_001014989.1 371 Missense Mutation GTC,TTC V43F NP_001014989.2
NM_014387.3 371 Missense Mutation GTC,TTC V7F NP_055202.1
Gene
SPNS1
Gene Name
sphingolipid transporter 1 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142448.1 371 Intron NP_001135920.1
NM_001142449.1 371 Intron NP_001135921.1
NM_001142450.1 371 Intron NP_001135922.1
NM_001142451.1 371 Intron NP_001135923.1
NM_032038.2 371 Intron NP_114427.1
XM_006721096.3 371 Intron XP_006721159.1
XM_017023757.1 371 Intron XP_016879246.1
XM_017023758.1 371 Intron XP_016879247.1
XM_017023759.1 371 Intron XP_016879248.1

View Full Product Details