Product Details

SNP ID
rs182050188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132747665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCGAAGCAACCGGAGCAGGTGCC[C/T]TGGGACCCGCCAGCCCGGAGCGGCC
Phenotype
MIM: 608418
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCNI2 PubMed Links

Gene Details

Gene
CCNI2
Gene Name
cyclin I family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039780.3 264 Missense Mutation CCT,CTT P57L NP_001034869.1
NM_001287252.1 264 Missense Mutation CCT,CTT P57L NP_001274181.1
NM_001287253.1 264 Missense Mutation CCT,CTT P57L NP_001274182.1
XM_017009725.1 264 Missense Mutation CCT,CTT P57L XP_016865214.1
Gene
SEPT8
Gene Name
septin 8
There are no transcripts associated with this gene.

View Full Product Details